siRNA Details
Gene Symbol: Pnpla6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122818.2 | NP_001116290.2 | 32 | 3898 | ||
NM_015801.2 | NP_056616.2 | 31 | 3851 | |||
GenBank mRNA | AF173829.1 | - | 31 | 3851 | ||
AK089095.1 | - | 28 | 3683 | |||
BC054789.1 | - | 32 | 3857 | 6831507 | ||
BC056999.1 | - | 32 | 3857 | 6831507 |

More Information
Other Species Targeted

Gene Ontology Categories:
