siRNA Details
Gene Symbol: Pnpla6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122818.2 | NP_001116290.2 | 28, 29 | 3368 | ||
NM_015801.2 | NP_056616.2 | 27, 28 | 3321 | |||
GenBank mRNA | AF173829.1 | - | 27, 28 | 3321 | ||
AK089095.1 | - | 25, 26 | 3270 | |||
AK162641.1 | - | 25, 26 | 3282 | |||
BC054789.1 | - | 28, 29 | 3327 | 6831507 | ||
BC056999.1 | - | 28, 29 | 3327 | 6831507 |

More Information
Other Species Targeted

Gene Ontology Categories:
