siRNA Details
Gene Symbol: Pnpla6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122818.2 | NP_001116290.2 | 15 | 1551 | ||
NM_015801.2 | NP_056616.2 | 14 | 1504 | |||
GenBank mRNA | AF173829.1 | - | 14 | 1504 | ||
AK040720.1 | - | 14 | 1962 | |||
AK089095.1 | - | 12 | 1453 | |||
AK162641.1 | - | 12 | 1465 | |||
AV582056.1 | - | 3 | 225 | |||
BC054789.1 | - | 15 | 1510 | 6831507 | ||
BC056999.1 | - | 15 | 1510 | 6831507 |
