siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 26 | 3065 | ||
NM_015805.3 | NP_056620.2 | 26 | 3065 | |||
GenBank mRNA | AF155913.1 | - | 25 | 2793 | ||
AK030485.1 | - | 25 | 2725 | |||
AK050115.1 | - | 5 | 468 | |||
BC003246.1 | - | 8 | 822 | |||
BC057000.1 | - | 25 | 2926 | |||
BC079626.1 | - | 26 | 2990 | 30536500 |
