siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 9, 10 | 1080 | ||
NM_015805.3 | NP_056620.2 | 9, 10 | 1080 | |||
GenBank mRNA | AB206122.1 | - | 8, 9 | 808 | ||
AF155913.1 | - | 8, 9 | 808 | |||
AK030485.1 | - | 8, 9 | 741 | |||
AK044331.1 | - | 9, 10 | 1055 | |||
BC057000.1 | - | 9, 10 | 1005 | |||
BC079626.1 | - | 9, 10 | 1005 | 30536500 |
