siRNA Details
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015806.5 | NP_056621.4 | 2 | 1090 | ||
NM_027418.2 | NP_081694.1 | 2 | 1094 | |||
GenBank mRNA | AF132850.1 | - | 1 | 823 | ||
AK044752.1 | - | 2 | 1072 | |||
BC006778.1 | - | 1 | 304 | |||
BC037060.1 | - | 1 | 828 | |||
BC048779.1 | - | 1 | 592 | 6315375 | ||
BC052420.1 | - | 1 | 339 | 5702905 | ||
BC100385.1 | - | 2 | 1072 | 30732053 |
