siRNA Details
Gene Symbol: Clec4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016751.3 | NP_058031.2 | 4 | 1027 | ||
GenBank mRNA | AK040696.1 | - | 4 | 1028 | ||
BC013647.1 | - | 4 | 987 | 4195561 | ||
D88577.1 | - | 4 | 943 |
