siRNA Details
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175238.5 | NP_780447.4 | 11, 12 | 1259 | ||
GenBank mRNA | AY428571.2 | - | 11, 12 | 1243 | ||
AY585206.1 | - | 10, 11 | 1252 | |||
JX255737.1 | - | 10, 11 | 1195 |
