siRNA Details
Gene Symbol: Ppp6r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164159.1 | NP_001157631.1 | 10 | 1338 | ||
NM_028999.1 | NP_083275.1 | 11 | 1486 | |||
NM_029456.2 | NP_083732.1 | 11 | 1486 | |||
GenBank mRNA | AK018652.1 | - | 10 | 1327 | ||
AK032179.1 | - | 2 | 257 | |||
AK035236.1 | - | 11 | 1466 | |||
AK088526.1 | - | 10 | 1306 | |||
AK129392.1 | - | 11 | 1428 | |||
BC008529.1 | - | 11 | 1469 | 3601021 |
