siRNA Details
Gene Symbol: Krr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178610.4 | NP_848725.2 | 2, 3 | 348 | ||
GenBank mRNA | AK011972.1 | - | 2, 3 | 258 | ||
AK030560.1 | - | 2, 3 | 260 | |||
AK032472.1 | - | 2, 3 | 260 | |||
AK036447.1 | - | 2, 3 | 269 | |||
AK041854.1 | - | 2, 3 | 260 | |||
AK146926.1 | - | 2, 3 | 266 | |||
AK168675.1 | - | 2, 3 | 260 | |||
BC089510.1 | - | 2, 3 | 265 | 30246541 | ||
BC094236.1 | - | 2, 3 | 249 | |||
CD547185.1 | - | 2, 3 | 348 |
