siRNA Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016985.2 | NP_058681.1 | 11 | 1530 | ||
GenBank mRNA | AF073997.1 | - | 11 | 1503 | ||
AK048714.1 | - | 3 | 783 | |||
AK051390.1 | - | 4 | 528 | |||
BC056376.1 | - | 11 | 1530 | 6849757 | ||
BC057337.1 | - | 11 | 1524 | 6825916 |
