siRNA Details
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016804.4 | NP_058084.3 | 5 | 433 | ||
GenBank mRNA | AB041562.1 | - | 5 | 425 | ||
AF053550.1 | - | 5 | 406 | |||
AK083055.1 | - | 395 | ||||
AK087933.1 | - | 5 | 421 | |||
AK168000.1 | - | 5 | 426 | |||
AK168021.1 | - | 5 | 396 | |||
AK168045.1 | - | 5 | 425 | |||
AK169048.1 | - | 5 | 429 | |||
AK179720.1 | - | 5 | 228 | |||
AK180612.1 | - | 2 | 54 | |||
AK208117.1 | - | 2 | 54 | |||
AK211256.1 | - | 2 | 54 | |||
AK214537.1 | - | 2 | 54 | |||
AK214538.1 | - | 2 | 54 | |||
AK218920.1 | - | 2 | 54 | |||
AK220102.1 | - | 2 | 54 | |||
BC006641.1 | - | 5 | 383 | 3483318 |
