siRNA Details
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098227.1 | NP_001091697.1 | 7 | 830 | ||
NM_016807.2 | NP_058087.2 | 7 | 827 | |||
GenBank mRNA | AF003693.1 | - | 7 | 822 | ||
AF077527.1 | - | 7 | 786 | |||
AK028429.1 | - | 7 | 803 | |||
AK033412.1 | - | 7 | 828 | |||
AK146085.1 | - | 7 | 802 | |||
AK150278.1 | - | 7 | 795 | |||
AK150321.1 | - | 7 | 798 | |||
AK150544.1 | - | 7 | 802 | |||
AK150623.1 | - | 7 | 806 | |||
AK150661.1 | - | 7 | 795 | |||
AK150707.1 | - | 7 | 801 | |||
AK150721.1 | - | 7 | 810 | |||
AK150736.1 | - | 7 | 795 | |||
AK150801.1 | - | 7 | 800 | |||
AK150996.1 | - | 7 | 802 | |||
AK151050.1 | - | 7 | 802 | |||
AK152001.1 | - | 7 | 801 | |||
AK152017.1 | - | 7 | 802 | |||
AK152080.1 | - | 7 | 810 | |||
AK152109.1 | - | 7 | 802 | |||
AK152434.1 | - | 7 | 802 | |||
AK152552.1 | - | 7 | 806 | |||
AK152577.1 | - | 7 | 801 | |||
AK152855.1 | - | 7 | 802 | |||
AK153042.1 | - | 7 | 802 | |||
AK153112.1 | - | 7 | 806 | |||
AK159124.1 | - | 7 | 798 | |||
AK159323.1 | - | 7 | 773 | |||
AK159538.1 | - | 7 | 800 | |||
AK159677.1 | - | 7 | 809 | |||
AK165657.1 | - | 7 | 797 | |||
AK167513.1 | - | 7 | 798 | |||
AK168004.1 | - | 7 | 805 | |||
AK168777.1 | - | 7 | 798 | |||
AK169219.1 | - | 7 | 807 | |||
AK169415.1 | - | 7 | 836 | |||
AK169687.1 | - | 7 | 802 | |||
AK171162.1 | - | 7 | 802 | |||
BC019400.1 | - | 7 | 793 | 4017442 |
