siRNA Details
Gene Symbol: Exoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162872.1 | NP_001156344.1 | 10 | 1315 | ||
NM_016857.2 | NP_058553.2 | 11 | 1408 | |||
GenBank mRNA | AF014461.1 | - | 11 | 1343 | ||
AK087601.1 | - | 10 | 1303 | |||
AK140450.1 | - | 11 | 1382 | |||
AK162868.1 | - | 10 | 1728 | |||
AK167769.1 | - | 10 | 1298 | |||
AK169928.1 | - | 10 | 1302 | |||
AK173094.1 | - | 9 | 5895 | |||
BC028927.1 | - | 10 | 1291 | 4208803 | ||
BC053710.1 | - | 10 | 1302 | 30056570 |
