siRNA Details
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017383.3 | NP_059079.2 | 22 | 3237 | ||
GenBank mRNA | AB032602.1 | - | 21 | 2923 | ||
AK052972.1 | - | 23 | 3130 | |||
BC076594.1 | - | 22 | 3237 | 30649661 |
