siRNA Details
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195083.1 | NP_001182012.1 | 2 | 459 | ||
NM_001195130.1 | NP_001182059.1 | 10 | 1859 | |||
NM_018774.4 | NP_061244.1 | 10 | 1852 | |||
GenBank mRNA | AB062362.1 | - | 2 | 445 | ||
AF060076.1 | - | 3 | 438 | |||
BC057571.1 | - | 10 | 1795 | 6410302 | ||
BC071246.1 | - | 2 | 440 | |||
U81491.1 | - | 9 | 1749 |
