siRNA Details
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018775.4 | NP_061245.3 | 7, 8 | 1404 | ||
GenBank mRNA | AK138630.1 | - | 7, 8 | 1402 | ||
AK141520.1 | - | 7, 8 | 1393 | |||
AK141816.1 | - | 7, 8 | 1358 | |||
AK162371.1 | - | 1, 2 | 179 | |||
BC005421.1 | - | 4, 5 | 739 | |||
BC065081.1 | - | 7, 8 | 1338 | 6832495 | ||
U88873.1 | - | 3, 4 | 624 |
