siRNA Details
Gene Symbol: Pde3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018779.1 | NP_061249.1 | 8 | 2288 | ||
GenBank mRNA | AF099187.1 | - | 8 | 2288 | ||
AK052374.1 | - | 8 | 1564 | |||
AK164864.1 | - | 8 | 1070 | |||
BC138265.1 | - | 8 | 2246 | 8861284 | ||
BC138266.1 | - | 8 | 2246 | 8861285 |
