siRNA Details
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001488.3 | NP_001001488.2 | 23, 24 | 3262 | ||
GenBank mRNA | AK157316.1 | - | 23, 24 | 3530 | ||
AY506548.1 | - | 23, 24 | 2991 | |||
BC117946.2 | - | 23, 24 | 2989 | 40098035 |
