siRNA Details
Gene Symbol: Slc1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018861.3 | NP_061349.3 | 8 | 1666 | ||
GenBank mRNA | AF246129.1 | - | 8 | 1425 | ||
AK044280.1 | - | 8 | 1637 | |||
AK136900.1 | - | 8 | 1666 | |||
AK139248.1 | - | 8 | 1665 | |||
AK140866.1 | - | 8 | 1667 | |||
AK148075.1 | - | 8 | 1667 | |||
BC043483.1 | - | 8 | 1624 | 5003744 | ||
BC052733.1 | - | 8 | 1540 | 6816689 | ||
U75215.1 | - | 8 | 1536 |
