siRNA Details
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018884.2 | NP_061372.2 | 10 | 2388 | ||
GenBank mRNA | AF127084.1 | - | 10 | 2388 | ||
AF127085.1 | - | 11 | 2232 | |||
AK143226.1 | - | 10 | 1902 | |||
AK173098.1 | - | 10 | 2347 | |||
BC010329.1 | - | 6 | 1208 | 3600603 | ||
HM560981.1 | - | 9 | 2280 |
