siRNA Details
Gene Symbol: Cpxm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018867.5 | NP_061355.3 | 2 | 501 | ||
GenBank mRNA | AF017639.1 | - | 2 | 502 | ||
AK019509.1 | - | 2 | 500 | |||
AK038874.1 | - | 2 | 384 | |||
BC021444.1 | - | 2 | 423 | 5038528 |
