siRNA Details
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019514.3 | NP_062387.2 | 17 | 3174 | ||
NM_207109.2 | NP_996992.1 | 18 | 3330 | |||
GenBank mRNA | AF116573.1 | - | 15 | 2342 | ||
AK041160.1 | - | 19 | 3234 |
