siRNA Details
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019514.3 | NP_062387.2 | 20, 21 | 3608 | ||
NM_207109.2 | NP_996992.1 | 21, 22 | 3764 | |||
GenBank mRNA | AF116573.1 | - | 18, 19 | 2776 | ||
AK041160.1 | - | 22, 23 | 3668 |
