siRNA Details
Gene Symbol: Pfpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019540.2 | NP_062413.2 | 2 | 1358 | ||
GenBank mRNA | AF250839.1 | - | 2 | 1334 | ||
AK141262.1 | - | 2 | 1292 | |||
BC052215.1 | - | 2 | 1326 | 30020114 |
