siRNA Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019795.4 | NP_062769.2 | 2 | 700 | ||
GenBank mRNA | AB028861.1 | - | 2 | 148 | ||
AK011875.1 | - | 2 | 139 | |||
AK076032.1 | - | 2 | 171 | |||
AK145506.1 | - | 2 | 162 | |||
AK145739.1 | - | 2 | 162 | |||
AK166872.1 | - | 2 | 162 | |||
AY323828.1 | - | 2 | 136 | |||
BC023681.1 | - | 2 | 344 | 5343384 | ||
BC055729.1 | - | 2 | 699 | 6414499 |
