siRNA Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019795.4 | NP_062769.2 | 9, 10 | 1562 | ||
GenBank mRNA | AB028861.1 | - | 9, 10 | 1010 | ||
AK011384.1 | - | 4, 5 | 420 | |||
AK011875.1 | - | 9, 10 | 1001 | |||
AK076032.1 | - | 9, 10 | 1033 | |||
AK145506.1 | - | 9, 10 | 1024 | |||
AK145739.1 | - | 9, 10 | 1024 | |||
AK166872.1 | - | 9, 10 | 1024 | |||
AY323828.1 | - | 9, 10 | 998 | |||
BC023681.1 | - | 9, 10 | 1206 | 5343384 | ||
BC055729.1 | - | 9, 10 | 1561 | 6414499 |
