siRNA Details
Gene Symbol: Sult1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019878.4 | NP_063931.1 | 8 | 945 | ||
GenBank mRNA | AF022894.1 | - | 8 | 964 | ||
AK033498.1 | - | 8 | 948 | |||
AK087595.1 | - | 8 | 981 | |||
BC024361.1 | - | 7 | 904 | 4990345 | ||
U92076.1 | - | 8 | 890 |
