siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 13 | 2474 | ||
NM_019831.3 | NP_062805.1 | 13 | 2334 | |||
GenBank mRNA | AF156605.1 | - | 13 | 2960 | ||
AK122268.1 | - | 11 | 2850 | |||
BC050131.1 | - | 13 | 2357 | |||
BC139001.1 | - | 12 | 2267 | 8862023 | ||
BC145497.1 | - | 12 | 2261 | 9054060 |
