siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 3 | 914 | ||
NM_001177986.1 | NP_001171457.1 | 3 | 914 | |||
NM_001177987.1 | NP_001171458.1 | 3 | 843 | |||
NM_001177988.1 | NP_001171459.1 | 3 | 761 | |||
NM_019831.3 | NP_062805.1 | 3 | 774 | |||
GenBank mRNA | AF156605.1 | - | 3 | 1400 | ||
AK033470.1 | - | 3 | 810 | |||
AK082075.1 | - | 3 | 757 | |||
AK122268.1 | - | 3 | 796 | |||
BC042699.1 | - | 3 | 779 | |||
BC050131.1 | - | 3 | 798 | |||
BC082320.1 | - | 3 | 843 | |||
BC108366.1 | - | 3 | 841 | |||
BC139001.1 | - | 2 | 707 | 8862023 | ||
BC145497.1 | - | 2 | 707 | 9054060 | ||
CB519077.1 | - | 3 | 759 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC106865.1 | - | 292 | Chr. - - [-] on Build |

More Information
Other Species Targeted

Gene Ontology Categories:
