siRNA Details
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019698.2 | NP_062672.2 | 19 | 2509 | ||
NM_153554.2 | NP_705782.2 | 19 | 2503 | |||
GenBank mRNA | AF056573.1 | - | 19 | 2414 | ||
AF056574.1 | - | 19 | 2408 | |||
AK151072.1 | - | 19 | 2431 | |||
AK153946.1 | - | 19 | 2430 | |||
AK159610.1 | - | 19 | 2431 | |||
AK166459.1 | - | 19 | 2395 | |||
AK166494.1 | - | 19 | 2396 | |||
AK168905.1 | - | 19 | 2429 | |||
AK189981.1 | - | 2 | 123 | |||
AK197048.1 | - | 3 | 332 | |||
AK197325.1 | - | 2 | 123 | |||
AK214351.1 | - | 2 | 123 | |||
AK215898.1 | - | 2 | 123 | |||
AK216599.1 | - | 2 | 123 | |||
BC033427.1 | - | 19 | 2440 | 5009115 | ||
BC037699.1 | - | 19 | 2388 | 4188900 |
