siRNA Details
Gene Symbol: Foxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019740.2 | NP_062714.1 | 3 | 1017 | ||
GenBank mRNA | AF114259.1 | - | 3 | 1017 | ||
AK047413.1 | - | 3 | 975 | |||
AK139629.1 | - | 3 | 1003 | |||
BC019532.1 | - | 2 | 857 |
