siRNA Details
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019680.2 | NP_062654.2 | 6 | 849 | ||
GenBank mRNA | AF016714.1 | - | 5 | 790 | ||
AK155203.1 | - | 6 | 1090 | |||
AK155709.1 | - | 6 | 850 | |||
BC109160.1 | - | 5 | 781 | 40047677 | ||
BC109171.1 | - | 5 | 781 | 40048316 |
