siRNA Details
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 10 | 1750 | ||
NM_019987.2 | NP_064371.2 | 10 | 1678 | |||
GenBank mRNA | AF225918.1 | - | 10 | 1518 | ||
AK052178.1 | - | 3 | 647 | |||
AK087484.1 | - | 10 | 1602 | |||
AK133537.1 | - | 10 | 1617 | |||
AK154185.1 | - | 10 | 1751 | |||
AK173068.1 | - | 5 | 907 | |||
BC028863.1 | - | 9 | 1435 | 4224269 |
