siRNA Details
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253709.1 | NP_001240638.1 | 7, 8 | 979 | ||
NM_001253713.1 | NP_001240642.1 | 8, 9 | 1087 | |||
NM_020007.3 | NP_064391.2 | 8, 9 | 2133 | |||
GenBank mRNA | AB093240.1 | - | 8, 9 | 2137 | ||
AK149405.1 | - | 2, 3 | 280 | |||
AK153827.1 | - | 7, 8 | 977 | |||
AK153848.1 | - | 7, 8 | 967 |
