siRNA Details
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253708.1 | NP_001240637.1 | 6, 7 | 2000 | ||
NM_001253709.1 | NP_001240638.1 | 5, 6 | 846 | |||
NM_001253710.1 | NP_001240639.1 | 5, 6 | 951 | |||
NM_001253713.1 | NP_001240642.1 | 6, 7 | 954 | |||
NM_020007.3 | NP_064391.2 | 6, 7 | 2000 | |||
GenBank mRNA | AB093240.1 | - | 6, 7 | 2004 | ||
AF231110.1 | - | 5, 6 | 1440 | |||
AK153827.1 | - | 5, 6 | 844 | |||
AK153848.1 | - | 5, 6 | 834 | |||
BC096600.1 | - | 5, 6 | 912 | 4976244 | ||
CN663545.1 | - | 3, 4 | 527 |
