siRNA Details
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205085.1 | NP_001192014.1 | 3 | 880 | ||
NM_020496.3 | NP_065242.1 | 3 | 880 | |||
NM_194263.2 | NP_919239.1 | 3 | 880 | |||
GenBank mRNA | AF260557.1 | - | 3 | 885 | ||
AF306667.1 | - | 3 | 818 | |||
AJ277486.1 | - | 3 | 422 | |||
AK052155.1 | - | 3 | 862 | |||
AK052389.1 | - | 3 | 863 | |||
AK141251.1 | - | 3 | 650 |
