siRNA Details
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205085.1 | NP_001192014.1 | 2 | 804 | ||
NM_020496.3 | NP_065242.1 | 2 | 804 | |||
NM_194263.2 | NP_919239.1 | 2 | 804 | |||
GenBank mRNA | AF306667.1 | - | 2 | 742 | ||
AJ277486.1 | - | 2 | 346 | |||
AK052155.1 | - | 2 | 786 | |||
AK052389.1 | - | 2 | 787 | |||
AK141251.1 | - | 2 | 574 |
