siRNA Details
Gene Symbol: Bcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020486.2 | NP_065232.1 | 13 | 1737 | ||
GenBank mRNA | AB035511.1 | - | 13 | 1698 | ||
AF109160.1 | - | 13 | 1737 | |||
AF221507.1 | - | 13 | 1687 | |||
AF346663.1 | - | 13 | 1675 | |||
AK075704.1 | - | 13 | 1702 | |||
AK158439.1 | - | 13 | 1703 | |||
BC004826.1 | - | 13 | 1721 | 2616148 |
