siRNA Details
Gene Symbol: Bcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020486.2 | NP_065232.1 | 2 | 226 | ||
GenBank mRNA | AB035511.1 | - | 2 | 187 | ||
AF221507.1 | - | 2 | 176 | |||
AF346663.1 | - | 2 | 164 | |||
AK040564.1 | - | 2 | 210 | |||
AK054001.1 | - | 2 | 207 | |||
AK075704.1 | - | 2 | 191 | |||
AK158439.1 | - | 2 | 186 | |||
BC004826.1 | - | 2 | 210 | 2616148 |
