siRNA Details
Gene Symbol: Stk32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162540.1 | NP_001156012.1 | 3 | 283 | ||
NM_021302.3 | NP_067277.2 | 3 | 583 | |||
GenBank mRNA | AB041542.1 | - | 3 | 563 | ||
AK046439.1 | - | 3 | 561 | |||
BC026457.1 | - | 3 | 568 | 4481218 |
