siRNA Details
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004468.2 | NP_001004468.1 | 7 | 5868 | ||
NM_021314.4 | NP_067289.2 | 3 | 781 | |||
NM_206856.3 | NP_996738.2 | 6 | 747 | |||
GenBank mRNA | AB041546.1 | - | 3 | 782 | ||
AK050985.1 | - | 4 | 676 | |||
AK145721.1 | - | 4 | 689 | |||
AK165194.1 | - | 3 | 721 | |||
AK165603.1 | - | 3 | 572 | |||
AY177410.1 | - | 5 | 759 |
