siRNA Details
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 13 | 1589 | ||
NM_001199276.2 | NP_001186205.1 | 11 | 1259 | |||
NM_001271455.1 | NP_001258384.1 | 12 | 1312 | |||
NM_001271456.1 | NP_001258385.1 | 13 | 1484 | |||
NM_021327.4 | NP_067302.2 | 12 | 1431 | |||
GenBank mRNA | AJ242777.1 | - | 11 | 1231 | ||
AJ242778.1 | - | 12 | 1403 | |||
AK156131.1 | - | 12 | 1285 | |||
AK170841.1 | - | 11 | 1237 | |||
AK172890.1 | - | 1217 | ||||
AK195559.1 | - | 2 | 156 | |||
AY012159.2 | - | 9 | 1001 | |||
BC008186.1 | - | 12 | 1348 | 3589311 | ||
BC008665.1 | - | 13 | 1455 | 3598934 | ||
CB599739.1 | - | 3 | 318 |
