siRNA Details
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178873.3 | NP_849204.1 | 2 | 1421 | ||
GenBank mRNA | AK028557.1 | - | 2 | 1425 | ||
AK028568.1 | - | 2 | 1422 | |||
AK161225.1 | - | 2 | 1424 | |||
BC069944.1 | - | 2 | 1169 | 30446117 |
