siRNA Details
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164197.1 | NP_001157669.1 | 9 | 1394 | ||
NM_021412.2 | NP_067387.1 | 9 | 1529 | |||
GenBank mRNA | AF153199.1 | - | 9 | 1481 | ||
AF155221.1 | - | 9 | 1509 | |||
AF162446.1 | - | 9 | 1509 | |||
AK079924.1 | - | 9 | 1395 | |||
AK080125.1 | - | 8 | 1532 | |||
AK086808.1 | - | 8 | 1269 | |||
AK136517.1 | - | 9 | 1383 | |||
AK154863.1 | - | 9 | 1523 | |||
BC112966.2 | - | 9 | 1501 | 40039023 |
