siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 2 | 304 | ||
GenBank mRNA | AF237814.1 | - | 2 | 114 | ||
AK009690.1 | - | 2 | 259 | |||
AK019106.1 | - | 2 | 266 | |||
AK031673.1 | - | 2 | 283 | |||
AK044905.1 | - | 2 | 265 | |||
AK049124.1 | - | 2 | 264 | |||
AK151549.1 | - | 2 | 258 | |||
AK159392.1 | - | 2 | 276 | |||
BC029205.1 | - | 2 | 221 | 4039560 | ||
BQ749205.1 | - | 2 | 277 | |||
CJ088919.1 | - | 2 | 304 |
