siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 26 | 4107 | ||
NM_021450.2 | NP_067425.2 | 26 | 4107 | |||
GenBank mRNA | AF149013.1 | - | 26 | 4057 | ||
AF376052.1 | - | 26 | 3963 | |||
AK029000.1 | - | 1 | 97 | |||
AK047673.1 | - | 3 | 1255 | |||
AY032951.1 | - | 26 | 4076 | |||
BC145122.1 | - | 26 | 3852 | |||
BC150709.1 | - | 26 | 3902 | 9087620 |
