siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 33 | 4972 | ||
NM_021450.2 | NP_067425.2 | 33 | 4975 | |||
GenBank mRNA | AF149013.1 | - | 33 | 4925 | ||
AF376052.1 | - | 33 | 4828 | |||
AK029000.1 | - | 8 | 962 | |||
AY032951.1 | - | 33 | 4944 | |||
BC026440.1 | - | 8 | 531 | |||
BC145122.1 | - | 33 | 4720 | |||
BC150709.1 | - | 33 | 4770 | 9087620 |
