siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 11 | 1451 | ||
GenBank mRNA | AF224508.1 | - | 11 | 1397 | ||
AK034237.1 | - | 11 | 1453 | |||
AK039404.1 | - | 11 | 1416 | |||
AK047863.1 | - | 4 | 386 | |||
AK171610.1 | - | 10 | 1227 | |||
AK173014.1 | - | 11 | 1285 | |||
BC030388.1 | - | 11 | 1413 | 5401143 |
