siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 18 | 2481 | ||
GenBank mRNA | AF224508.1 | - | 18 | 2427 | ||
AK034237.1 | - | 18 | 2483 | |||
AK039404.1 | - | 18 | 2446 | |||
AK047863.1 | - | 11 | 1415 | |||
AK171610.1 | - | 17 | 2257 | |||
AK173014.1 | - | 18 | 2315 | |||
AK183206.1 | - | 3 | 227 | |||
BC030388.1 | - | 18 | 2443 | 5401143 |
